Human SAPAP1 Peptide (14 aa in the center)
89419-754
: Prosci
:
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:SAPAP1 Peptide (14 aa in the center)
- Tested applications:Blocking
- Cat. no.:89419-754
- Supplier No.:4623P
SAPAP1 peptide is used for blocking the activity of SAPAP1 antibody.