Mouse SLPI Peptide (17 aa in the center)
Catalog # 89419-454
Supplier: Prosci
CAS Number:
Specifications
- Species:Mouse
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa in the center
- Protein/peptide name:SLPI Peptide (17 aa in the center)
- Tested applications:Blocking
- Cat. no.:89419-454
- Supplier No.:4251P
Specifications
About this item
SLPI peptide is used for blocking the activity of SLPI antibody.