Avian Influenza A (H5N1) Neuraminidase Influenza A Peptide (15 AA Near C-Terminus)
89418-932
: Prosci
:
- Species:Avian influenza A (H5N1) neuraminidase
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:Influenza A Peptide (15 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89418-932
- Supplier No.:3423P
The peptide is used for blocking the activity of the avian influenza neuraminidase antibody.