ContactUsLinkComponent
Granite Balance Table, Rutland Marble & Granite
Granite Balance Table, Rutland Marble & Granite
Catalog # 12568-012
Small Business Enterprise
Granite Balance Table, Rutland Marble & Granite
Catalog # 12568-012
Supplier Number:  BT144G
Small Business Enterprise
Specifications
  • Description:
    Grey Granite Balance Table
  • Height:
    31"
  • Environmentally Preferable:
  • Cat. no.:
    12568-012
  • W×D:
    35×24"
  • Supplier No.:
    BT144G
Specifications
About this item
This granite balance table is constructed of carefully crafted and sturdy 7.6 cm (3") thick stone slabs with vibration dampening mounts.
  • Balance tables are designed for use in laboratories, schools, and technical development centers
Built to last stone balance table legs are reinforced with a rigid stabilizing bar.