Jump to:
Product Details & Documents
- CAS Number:79-27-6
- Formula:Br₂HCCHBr₂
- MDL Number:MFCD00000133
- Molecular Weight:345.65 g/mol
- Merck Index:13,09261
- Boiling Pt:244 °C (1013 hPa)
- Melting Pt:–1 °C
- Density:2.9656 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-191-5
- UN:2504
- ADR:6.1,III
Documents
spec_CA71009-074.pdf
Product specification
Specifications
Specifications
- Pack type:Glass, amber
- Size:3 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



