D-(+)-Sucrose, crystals ACS
Product CAJT4072-09 is similar to CA71008-918 but manufacturer and specifications may vary.
Product Details & Documents
- CAS Number:57-50-1
- Formula:C₁₂H₂₂O₁₁
- MDL Number:MFCD00006626
- Molecular Weight:342.3 g/mol
- Merck Index:13,08966
- Melting Pt:169…170 °C
- Density:1.29 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-334-9
Documents
Specifications
Specification Test Results
- Specific rotation (10% solution.):+66.3° to +66.8°
- Insoluble Matter:0.005% max.
- Loss on drying at 105°C:0.03% max.
- Residue after ignition:0.01% max.
- Titrable acid:0.0008 meq/g max.
- Chloride (Cl):0.005% max.
- Sulfate and sulfite (as SO4):0.005% max.
- Heavy metals (as Pb):5 ppm max.
- Iron (Fe):5 ppm max.
- Invert Sugar:0.05% max.
Specifications
- Pack type:Poly pail
- Size:10 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



