You searched for: Furnaces (Ashing and Muffle Ovens)
Ashing and muffle ovens accurately heat to the selected temperature quickly improving energy efficiency and process times. The digital temperature controls ensure precise furnace heating level. With strong insulated construction, heat is distributed evenly while allowing exterior to stay cool. Special safety features include immediate power shutoff when door opens, over-temperature protection settings, and an adjustable alarm provide maximum user protection. The maximum temperature, dimensions, and control type varies among furnaces.
Lindberg/Blue M Box Furnaces, 1100 °C, Thermo Scientific
Supplier: Thermo Fisher Scientific
Small, reliable furnaces meet the specific needs of schools, as well as chemical and industrial laboratories
Expand 6 Items
Cupellation Muffle Furnace, 50-Place , Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Available with natural gas, liquefied petroleum gas, or oil (diesel) fuel source options
Expand 2 Items
168-Place Cupellation Furnace, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
These 168-place cupellation furnaces are available with LPG, natural gas, or diesel fuel source options. Cupels should be 168 x 6A, 7A, or 7AS. Optional port viewing window, rear extraction unit and hand valve door control extras are also available.
Expand 5 Items
Barnstead/Thermolyne Compact Benchtop Muffle Furnaces, Type 1500, Thermo Scientific
Supplier: Thermo Fisher Scientific
Economical, versatile furnaces have heavy-duty firebrick insulation surrounding the chamber opening for added durability
Expand 2 Items
Cupellation Muffle Furnace, 100-Place, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Available with natural gas, liquefied petroleum gas, or oil (diesel) fuel source options
Expand 5 Items
25/42 Place Fusion Furnaces, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
These 25/42-place fusion furnaces are available with LPG, natural gas or diesel fuel source options. Crucibles should be 25 x 50g or 42 x 40g.
Expand 5 Items
50/84 Place Fusion Furnace, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
These 50/84-place fusion furnaces are available with LPG, natural gas, or diesel fuel source options
Expand 5 Items
MAS-3 Furnaces, Anachemia
Supplier: A.R. Olympique
The Mine Assay MAS-3 fire assay furnace, formerly known as “Williams & Wilson,” is well known in the mining industry all across North America, as well as in other locations worldwide.
Expand 8 Items
Barnstead/Thermolyne Ashing Furnaces, Type 6000 and Type 30400, Thermo Scientific
Supplier: Thermo Fisher Scientific
Designed for coal and coke analysis procedures.
Expand 5 Items
Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Supplier: Thermo Fisher Scientific
Furnaces are designed for continuous operation to 1200 °C maximum and feature an integral controller in a single space-saving cabinet.
Expand 3 Items
Barnstead/Thermolyne Small Benchtop Muffle Furnaces, Type 1300 and Type 1400, Thermo Scientific
Supplier: Thermo Fisher Scientific
Versatile and economical Barnstead/Thermolyne furnaces are specifically designed for ashing organic and inorganic samples, determining volatile and suspended solids, gravimetric analysis, heat treating of small steel parts, and ignition tests.
Expand 4 Items
Thermolyne™ Benchtop Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
Furnaces ideal for gravimetric analysis, ashing of organic and inorganic samples, sintering, quantitative analysis and heat treating.
Expand 2 Items
Thermolyne™ Premium Large Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
Furnaces for fusions, brazing, enameling, drying precipitates, and volatile solids analysis.
Expand 8 Items
Barnstead/Thermolyne Benchtop Muffle Furnaces, Type 47900 and Type 48000, Thermo Scientific
Supplier: Thermo Fisher Scientific
Compact benchtop muffle furnaces feature advanced electronic control
Expand 5 Items
Barnstead/Thermolyne Large Chamber Muffle Furnaces, Type 30400, Thermo Scientific
Supplier: Thermo Fisher Scientific
Ideal for annealing glass, determining volatiles, ashing pulp and paper samples, and for catalyst research
Expand 5 Items
Melting Furnaces, Nabertherm
Supplier: NABERTHERM INC.
These compact melting furnaces for the melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages
Expand 13 Items
High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm
Supplier: NABERTHERM INC.
Designed as tabletop models, these compact high-temperature furnaces have a variety of advantages.
Expand 16 Items
Thermolyne™ Industrial Benchtop Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
Furnaces ideal for gravimetric analysis, heat treatment
Expand 2 Items
Muffle Furnaces up to 1200 °C, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
These muffle furnaces have been proven for daily laboratory use. These models stand out for their workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a fold down or lift up door at no extra charge.
Expand 45 Items
Heat Treating Furnace, Model AC1228/PM6, Cress
Supplier: CRESS MFG CO INC
Model AC1228/PM6 is a heat treating furnace used for treating metals with a maximum temperature of up to 2250°F.
Expand 1 Items
Chamber Furnaces for Annealing, Hardening and Brazing, Nabertherm
Supplier: NABERTHERM INC.
The N 7/H - N 87/H chamber furnaces are designed to withstand harsh use in the laboratory.
Expand 19 Items
High-Temperature Ovens, Forced Convection Chamber Furnaces, Models N and NA, Nabertherm
Supplier: NABERTHERM INC.
These chamber furnaces with air circulation are characterized by their extremely high temperature uniformity.
Expand 31 Items
Muffle Furnaces with Scale and Software, Tmax 1200 °C, L 9/12 SW or LT 9/12 SW, Nabertherm
Supplier: NABERTHERM INC.
These weighing furnaces with integrated precision scale and software, were designed especially for determining combustion loss in the laboratory. The determination of combustion loss is necessary, for instance, when analyzing sludges. Combustion loss is defined as the difference between the initial total mass and the combustion residue. During the process the software supplied records both temperature and weight loss.
Expand 8 Items
Muffle Furnaces up to 1100 °C, Nabertherm
Supplier: NABERTHERM INC.
With their performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the laboratory. Quality features such as the dual shell furnace housing of rust-free stainless steel, their compact, lightweight construction, or the heating elements encased in quartz glass tubes make these models reliable partners for your application.
Expand 9 Items
High-Temperature Furnaces with MoSi₂ Heating Elements, Fiber Insulation up to 1800 °C, Model HT, Nabertherm
Supplier: NABERTHERM INC.
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed.
Expand 7 Items
Muffle Furnaces with Embedded Heating Elements in the Ceramic Muffle, Nabertherm
Supplier: NABERTHERM INC.
The L 9/11/SKM muffle furnaces are particularly recommended for applications that involve aggressive substances.
Expand 8 Items
Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm
Supplier: NABERTHERM INC.
The ashing furnaces LV 3/11 - LVT 15/11 are especially designed for ashing in the laboratory.
Expand 35 Items
High Performance Muffle Furnace, FP Series, Yamato
Supplier: Yamato Scientific
Muffle furnace is designed to maintain uniform high heat temperatures and make the unit last for years. The units in this line have firmly sealed chamber doors to provide excellent heat tightness. They also come with a top-quality alumina porcelain hot plate where the heater is not exposed to the inner chamber. This ensures that no sample contamination will take place.
Expand 3 Items
Muffle Furnaces with Brick Insulation, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
Heating elements on support tubes radiating freely into the furnace chamber provide for particularly short heating times for these muffle furnaces.
Expand 20 Items
High-Temperature Furnaces with SiC Rod Heating, Up to 1600 °C, Nabertherm
Supplier: NABERTHERM INC.
These powerful laboratory muffle furnaces are available for temperatures up to 1400, 1500, 1550 or 1600 °C.