Order Entry
Canada
ContactUsLinkComponent
 
 
 
 
Potassium persulfate ≥99.0%, white crystalline powder ACS
 :  Avantor
 
 
 
 
Potassium persulfate ≥99.0%, white crystalline powder ACS
 :  Avantor
 

 

  • Assay :
    ≥ 99.0 %
  • Chlorine compounds (as Cl) :
    ≤ 0.001 %
  • Fe (Iron) :
    ≤ 5 PPM
  • Heavy Metals (as Pb) :
    ≤ 0.001 %
  • Insoluble Matter :
    ≤ 0.005 %
  • Mn (Manganese) :
    ≤ 2 PPM

 

Frequently Bought Together