Hydrochloric acid 0.5 N volumetric solution, traceable to NIST Standard Reference Material (SRM)
Supplier: VWR
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:36.46
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Normality:0.495 - .0505 N
Specification Test Results
Related Information
Frequently Bought Together