
Citric acid monohydrate 99.0-102.0%, granules ACS
Supplier: VWR
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (C6H8O7.H2O):99.0 - 102%
- Insoluble Matter:0.005% max.
- Residue after ignition:0.02% max.
- Chloride (Cl):0.001% max.
- Oxalate (C2O4):Passes Test
- Phosphate (PO4):0.001% max.
- Sulfate (SO4):0.002% max.
- Iron (Fe):3 ppm max.
- Lead (Pb):2 ppm max.
- Substances carbonizable by hot H2SO5:Passes Test
- Appearance:White granules
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_ANAC25680-380.pdf spec_CA71006-948 spec_CA71006-952
Product specification
Frequently Bought Together