Order Entry
Canada
Orders LinkContactUsLinkComponent
Isoamyl alcohol ≥98.5% ACS

Isoamyl alcohol ≥98.5% ACS

Glass, amber, 4 L
Catalog # CA71006-632
Flammable
Irritant
Isoamyl alcohol ≥98.5% ACS
Catalog # CA71006-632
Supplier:  
CAS Number:  123-51-3
MOC:  Non-RegulatedCertificates
Isoamyl alcohol ≥98.5% ACS
Isoamyl alcohol ≥98.5% ACS

Glass, amber, 4 L

CAS Number:  123-51-3
MOC:  Non-Regulated
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ISO-AMYL ALCOHOL 4L Catalog #CAAX1440-3

Product CAAX1440-3 is similar to CA71006-632 but manufacturer and specifications may vary.

3-METHYL-1-BUTANOL, ACS 4L Catalog #CAAA36716-K7

Product CAAA36716-K7 is similar to CA71006-632 but manufacturer and specifications may vary.

ISOAMYL ALCOHOL ULTRA PURE GRADE 1L Catalog #97065-028

Product 97065-028 is similar to CA71006-632 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
  • CAS Number:123-51-3
  • Formula:(CH₃)₂CHCH₂CH₂OH
  • MDL Number:MFCD00002934
  • Molecular Weight:88.15 g/mol
  • Merck Index:13,05214
  • Boiling Pt:130 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.812 g/cm³ (20 °C)
  • Flash Pt:42 °C
  • Storage Temperature:Ambient
  • EINECS:204-633-5
  • UN:1105
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Assay (C5H11OH):
    98.5% min.
  • Water (H2O):
    0.5% max.
  • Titrable acid:
    0.002 meq/g max.
  • Residue after evaporation:
    0.003% max.
  • Acid and esters (as amyl acetate):
    0.2% max.
  • Carbonyl compounds (as HCHO):
    0.1% max.

Specifications

  • Pack type:
    Glass, amber
  • Size:
    4 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.