Flip-Lok™ Vial, Empty
Product Details & Documents
Flip-Lok™ Vial, empty, sterile, polypropylene, clarified polypropylene, with hinged cap, large write-on area, high profile, 145 ml total volume, 250/case.
- Polypropylene is 100% recyclable
- Storage temperature: 15 to 30 °C
This highly versatile container can meet your needs. Manufactured by Hardy Diagnostics, the new Flip-Lok™ vial is suited for a wide variety of applications. Great for sample collection, and sample storage. Flip-Lid allows for easy one-handed opening. Sterile when unopened, this vial ensures product or sample integrity.
Vials are made of sturdy polypropylene with a leak-proof seal that holds up even during air transport. Wide 45 mm opening accommodates bulky samples. Also features plenty of head space for large samples. Each vial is clearly marked with a 90 and 99 ml fill line. Flip-top lid allows for easy one-handed opening and closing to make pipeting easier. Saves unnecessary motion! No awkward screw caps or plastic strips to tear off.
Manufactured by Hardy Diagnostics, the new Flip-Lok™ vial is suited for a wide variety of applications. Great for sample collection, and sample storage. Flip-Lid allows for easy one-handed opening. Sterile when unopened, this vial ensures product or sample integrity.
Certifications: Sterile
Ordering information: Also available filed with peptone water, buffered peptone water, butterfield's buffer, deionized water, and phosphate buffer with MgCl.
Documents
Specifications
- Cap included:Yes
- Sterility:Sterile
- Capacity:145.00 ml
- Color:Clear
- Material:Polypropylene
- Packaging:250/case
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



