ContactUsLinkComponent
Simple Machines, Gear Rack Model
Simple Machines, Gear Rack Model
Catalog # 470210-816
Supplier:  Wards
undefined
Simple Machines, Gear Rack Model
Catalog # 470210-816
Supplier:  Wards
Supplier Number:  WDMS15
Specifications
  • Description:
    Simple Machines, Gear Rack Model
  • Cat. no.:
    470210-816
  • Supplier No.:
    WDMS15
Specifications
About this item
This wooden machine has a sturdy wooden construction ideal for classroom use. This apparatus can demonstrate how gears can turn rotational motion into translational motion.
Includes an instruction manual to facilitate student and teacher use of the apparatus.