Simple Machines, Pendulum Model
Catalog # 470210-812
Supplier: Wards
Specifications
- Description:Simple Machines, Pendulum Model
- Cat. no.:470210-812
- Supplier No.:WDMS13
Specifications
About this item
This wooden pendulum has a strong and sturdy construction ideal for classroom use.
- Adjustable pendulum
- Two-dimensional movement
- Includes manual
The blue pendulum bob is adjustable so that students can easily and quantitatively study the relationship between the length of the pendulum and the period of the bob. Wooden guide helps students to keep the release angle uniform and small and the rigid body allows for movement in only two dimensions keeping students focused on the relationship of length and period without having to worry about other variables. Includes an instruction manual to facilitate student and teacher use of the apparatus.