Order Entry
Canada
Orders LinkContactUsLinkComponent
Fluid K

Fluid K

Fluid K, USP, 10/pack, 300 ml
Catalog # CA89495-512
Supplier #:  U348
Fluid K
Catalog # CA89495-512
CAS Number:  
Fluid K
Fluid K

Fluid K, USP, 10/pack, 300 ml

CAS Number:  
Price
$1,540.00
Log in for availability
Pack of 10 (300ml)
Minimum order quantity is 1
Product Details & Documents

Fluid K, USP, 300 ml fill, 500 ml glass bottle with needle-port septum screw cap, order by the package of 10.

Sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.

USP Fluids A, D, and K are used by the pharmaceutical and medical device industry in procedures used to ensure sterility of injectables, non-injectables, liquid products, oils and ointments, antibiotic powders, sterile pathway devices, and aerosols.

Fluid K is used for diluting, neutralizing, dissolving, and rinsing of devices and membrane filters according to USP procedures.

Certifications: USP

Specifications

Specifications

  • Description:
    Fluid K, USP
  • Size:
    10/pack
  • Fill volume:
    300 ml
  • Cat. No.:
    CA89495-512
  • Storage temperature:
    2-8C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.