Order Entry
Canada
ContactUsLinkComponent
Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics
Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics
Catalog # CA52428-868
undefined
Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics
Catalog # CA52428-868
Supplier Number:  AB5819

Specifications

  • Description:
    Autoclave bag
  • Size:
    8×12"
  • Cat. no.:
    CA52428-868
  • Packaging:
    BAGA
  • Supplier No.:
    AB5819

Specifications

About this item

Autoclave bag is made of tough, thick polypropylene, double seamed bottom, solid red bag with black biohazard warnings in English and Spanish.

  • Dimension: 8×12"
  • Thickness: 2 mil
  • Bag is rated to withstand temperatures up to 250 °F

Storage temperature: 15...30 °C. Heat indicator strip turns black when exposed to steam, OSHA approved labeling, plastic twist ties for closure furnished upon request.

Packaging: 400 per case