ContactUsLinkComponent
Visual Scientifics Wave Demonstration
Visual Scientifics Wave Demonstration
Catalog # 470212-494
Supplier:  Wards
undefined
Visual Scientifics Wave Demonstration
Catalog # 470212-494
Supplier:  Wards
Supplier Number:  PTWDA
Specifications
  • Description:
    Visual scientifics wave demonstration
  • Cat. no.:
    470212-494
  • Supplier No.:
    PTWDA
Specifications
About this item
Students learn how the length, tension, and mass of a string affect the frequency of a standing wave in this engaging demo.
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
  • 10+ page instruction book included
This apparatus is designed to guide students through understanding standing waves, nodes and antinodes, period and frequency, and tension. The included instruction booklet provides approximately three hours of instructional activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts.
Ordering information: Visual Scientifics Base (470148-822) and magnetic ring stands are required, but sold separately.
Delivery: This kit includes thin and thick string, an oscillating motor, and a mountable pulley. Required but not included: Ring stand posts, video camera and graphical analysis software, and a digital balance.