Culture Tube Cap, Hach
Supplier: Hach
Certificates
About this item
White cap for Culture Tube
Screw cap has a firmly bonded fluoropolymer liner, providing a sure and highly inert sealing surface. Package of 6.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.