Model HA-4P MG-L Hardness (Total and Calcium) Test Kit, Hach
Supplier: Hach
Certificates
About this item
Model HA-4P MG-L is a hardness, total and calcium (magnesium by difference) drop count titration/EDTA test kit with a range of 20-400 mg/L
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Contains 100 tests. This kit utilizes drop-count titration and is simple and economical. Contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Ordering information: Kit includes five reagents, bottle, measuring tube, instruction sheet, and carrying case.
Frequently Bought Together