Model ASC-1 Ascorbic Acid Test Kit, Hach
Model ASC-1 Ascorbic Acid 100-Test Kit was developed in cooperation with beverage producers and uses the drop-count titration method to measure ascorbic acid (vitamin C)
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Utilizes drop count titration with a range of 10-200 mg/L. This kit utilizes drop-count titration simply and economically. It contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
: Sample measuring tube, flask, reagents, instructions and carrying case included.
Model ASC-1 Ascorbic Acid 100-Test Kit was developed in cooperation with beverage producers and uses the drop-count titration method to measure ascorbic acid (vitamin C)
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Utilizes drop count titration with a range of 10-200 mg/L. This kit utilizes drop-count titration simply and economically. It contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
: Sample measuring tube, flask, reagents, instructions and carrying case included.
- Catalog No:
- 89500-718
- CA89500-718
- Method:
- Drop Count Titration/ Iodometric
- Drop Count Titration/ Iodometric
- Platform:
- DCT
- DCT
Product Family Options
- Chemical methodRangeNo. of tests
- Iodometric10 - 200 mg/L ascorbic acid100-: -Product available on GSA Advantage®
Specifications
Cat. No.89500-718MethodDrop Count Titration/ IodometricPlatformDCT - : 2308100
Specifications
Cat. No.CA89500-718MethodDrop Count Titration/ IodometricPlatformDCT