Chromium Test Kit, Color Disc Tests, Model CH-12, Hach
: Hach
Widely used as a corrosion inhibitor in cooling towers and engine cooling systems, chromium is also used in surface finishing, leather tanning, and other industries. The most widely used form, hexavalent chromium, is very toxic and strict limits are placed on discharges. Method/Chemistry: Color disc. Smallest Increment: 0.1 mg/L. Case Style: 12 x 7.25x 4.5". Blue Polypropylene Carrying Case.
Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the lab or field.
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
: Four reagents, clippers, two color discs, comparator box, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, instructions and carrying case included.
- Method:Color disc/ Diphenylcarbazide, Hypobromite Oxidation
Frequently Bought Together