Model HA-4P Hardness (Total and Calcium) Test Kit, Hach
: Hach
Hardness, Total & Calcium (as CaCO3) drop-count titration/EDTA 100-test kit with range of 1-20 gpg
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
This test kit utilizes a simple and economical method. It contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
: Kit includes five reagents, bottle, measuring tube, instruction sheet, and carrying case.
- Method:Drop Count Titration / EDTA
- Platform:DCT
Frequently Bought Together