Sodium thiosulfate 0.113 N, cartridge for digital titrator
Catalog # CA97026-432
Supplier: Hach
CAS Number:
Specifications
- Size:1 cartridge
- Cat. no.:CA97026-432
- Supplier No.:2267301
Specifications
Specification Test Results
- Method:8209
- Method Name:Iodometric
- Number of tests:40 - 100
- Parameter:Chlorine, Total
- Platform :Digital Titrator
- Range:20 - 2000 mg/L Cl2
- Titrant:0.113 N Na2S2O3
Specification Test Results
About this item
:
:158.11 g/mol
Storage Temperature:Ambient
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499