Sodium thiosulfate 2.00 N, cartridge for digital titrator
CA97026-242
: Hach
:
- Size:1 cartridge
- Cat. no.:CA97026-242
- Supplier No.:1440101
- Method:8209
- undefined:Azide mod. of Winkler Method
- Method Name:Iodometric
- Number of tests:40 - 100
- Parameter:Chlorine, Total
- Platform :Digital Titrator
- Range:2000 - 70000 mg/L Cl2
- Range 2:10 - 100 mg/L DO
- Titrant:2.00 N Na2S2O3
:
:158.11 g/mol
:Ambient
:7772-98-7
:Na₂S₂O₃
:MFCD00003499