
Sodium hydroxide 3.636 N, cartridge for digital titrator
Catalog # CA14380-01
Supplier: Hach
CAS Number:
Specifications
- Size:1 cartridge
- Cat. no.:CA14380-01
- Supplier No.:14380-01
Specifications
Specification Test Results
- Method:8205
- Method Name:Phenolphthalein
- Number of tests:40 - 100
- Parameter:Carbon Dioxide
- Platform :Digital Titrator
- Range:100 - 1000 mg/L CO2
- Titrant:3.636 N NaOH
Specification Test Results
About this item
:
:40 g/mol
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548