ContactUsLinkComponent
Starfish Gas/Vacuum-Distribution Manifold with Connector
Starfish Gas/Vacuum-Distribution Manifold with Connector
  CA11009-082
Starfish Gas/Vacuum-Distribution Manifold with Connector
  CA11009-082
 :  015890350CA
  • Description:
    Gas/vacuum-distribution manifold with connector
  • Cat. no.:
    CA11009-082
  • Supplier No.:
    015890350CA
Gas/Vacuum distribution manifold allows gas or vacuum from a single source to be evenly distributed to up to five positions or vessels (does not control or regulate gas/vacuum flow).
Connectors feature leak-proof shut-off valves.