Specifications
- Description:SlideRite™ Water Soluble Spray
- Cat. no.:CA11027-246
- Supplier No.:1801
Specifications
About this item
Slide Rite is a highly concentrated, water-soluble spray that forms a lasting protective coating over all types of cytological smears
Many cytological specimens are fragile and degenerate quickly. Therefore an immediate fixation of the material to be studied is essential.