SP Bel-Art Three-Way Fluoroplastic Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
The three-way configuration of these stopcocks permits connection of any two or all three ports.
- PVDF construction
- Chemically resistant
- Autoclavable
Barbed tubulations accept rubber or plastic tubing. They are made of PVDF (polyvinylidene fluoride), a fluoroplastic resistant to most chemicals that withstands temperatures up to 204ºC (400ºF). Maximum pressure rating 34.3 KPa (5 psi). Steam autoclavable.
Related Information
Certificates
Search by Lot NumberWilmad LabGlass Media Formulation and Transfer for Vaccine Development
Product Literature
Frequently Bought Together