DOSE IT Peristaltic Pump, INTEGRA Biosciences
Catalog #: 89498-812
Supplier: INTEGRA Biosciences
Specifications
- Max. flow rate:5 L/min
- Min. flow rate:0.6 ml/min
- Description:DOSE IT Peristaltic Pump
- Electrical:100–240VAC 50/60Hz
- Dosing volume:0.1 mL – 9999 mL
- Interfaces:2x RS232
- Int. Ø tubing:1 - 8 mm
- Weight:3.5 kg (7.7 lb)
- Cat. no.:89498-812
- Wall Thickness:1.5 mm (1/16"), tubing wall
- Dimensions:203H x 210W x 191D mm (8.0 x 8.27 x 7.51")
- Supplier No.:171000
Specifications
About this item
The peristaltic pump DOSE IT makes dispensing of culture media, buffers and other solutions easy and efficient
With its very small footprint and light weight, the DOSE IT fits everywhere in the lab and can be moved easily–no need to waste valuable bench space or to have a pump in every lab.
In addition to the dispensing and pump mode, the DOSE IT provides the ability to create customized liquid handling protocols. Easy to follow instructions on the display allow users to set up individual protocols, consisting of consecutive application steps.
Certifications: CE certified.
Ordering information: Includes 1 silicone tubing, inner diameter 4 mm, length 2.5 m, 2 aspiration/dispensing tubes, length 10 cm, stainless steel, one end dented, 1 aspiration/dispensing tube, length 35 cm stainless steel, one end dented, 1 tube collar, 1 pipette-tubing connector and mains adapter.
Customers Who Bought This Also Bought