Wide Mouth Glass Powder Funnel with Duran Flange, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This is a borosilicate glass wide mouth funnel with a Duran® flange bottom
Flange matches side ports on Ace Glass reactor heads. Funnel is slightly angled to allow for easy pouring of material into large scale reactors. Uses quick-release clamps.
Customers Who Bought This Also Bought