Glutaraldehyde 25% in water for synthesis, Sigma-Aldrich®
Fournisseur: MilliporeSigma
Certificats
À propos de cet article
Numéro CAS:111-30-8
:
Formule:C₅H₈O₂
Numéro MDL:MFCD00007025
:100.12
MARC:8 (6.1),II
:M-Clarity™ quality level = MQ200
ONU:2922
:GHS05
:GHS08
:GHS07
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Résultats des tests de spécification
- Solubility:(20 °C) soluble
- Melting Point:-6 °C
- Boiling Point:100.5 °C (1013 hPa)
- Vapor Pressure:0.27 hPa (20 °C)
- Density:1.06 g/cm3 (20 °C)
- pH:3.0 (H2O, 20 °C)
- Ignition Point:275 °C
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Résultats des tests de spécification
Informations connexes
Frequently Bought Together