

Deuterium chloride 20% in D₂O (99.5% D) (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:7698-05-7
:
Formula:DCl
MDL Number:MFCD00044642
:37.45
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:1789
:GHS05
:GHS07
Specification Test Results
- Density:1.013 g/cm3 (20 °C)
- pH:0.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot Number12146085.pdf
SDS
AYN4_Analytical_Chemistry_-_Spectroscopy
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together