Order Entry
Canada
ContactUsLinkComponent
 
Vanadium Standard, Titrisol®, MilliporeSigma
Catalog # CA1.09994.0001
Supplier:  MilliporeSigma
CAS Number:  
Vanadium Standard, Titrisol®, MilliporeSigma
Catalog # CA1.09994.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09994.0001
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    8,6% H₂SO₄
  • Pack type:
    Plastic Ampule
  • Size:
    1 Ampule
  • Cat. no.:
    CA1.09994.0001
  • Element:
    Vanadium
  • Supplier No.:
    1.09994.0001

Specifications

About this item