Order Entry
Canada
ContactUsLinkComponent
Hydrochloric acid 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Corrosive
Hydrochloric acid 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09974.0001
Supplier:  MilliporeSigma
CAS Number:  
Corrosive
Hydrochloric acid 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09974.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09974.0001
CAS Number:  

Specifications

  • Pack type:
    Cardboard box
  • Size:
    1 piece
  • Cat. no.:
    CA1.09974.0001
  • Supplier No.:
    1.09974.0001

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    1 (H2O) (undiluted)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,
 :36.46
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324