Sodium thiosulfate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09950.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Cardboard box
- Size:1 pc.
- Cat. no.:CA1.09950.0001
- Supplier No.:1.09950.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.22 g/cm3 (20 °C)
- pH:9 - 10 (H2O, 20 °C)
Specification Test Results
About this item
:
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499