Order Entry
Canada
ContactUsLinkComponent
Sodium carbonate 0.1 N concentrated aqueous solution, Titrisol®, Supelco®
Sodium carbonate 0.1 N concentrated aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09940.0001
Supplier:  MilliporeSigma
CAS Number:  
undefined
Sodium carbonate 0.1 N concentrated aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09940.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09940.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Cardboard box
  • Size:
    1 pc.
  • Environmentally Preferable:
  • Cat. no.:
    CA1.09940.0001
  • Supplier No.:
    1.09940.0001

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.1 g/cm3 (20 °C)
  • pH:
    11.6 (H2O, 20 °C)

Specification Test Results

About this item

Synonyms: Calcined soda, Carbonic acid disodium salt
 :
 :105.99 g/mol
Boiling Pt:1600
Melting Pt:854
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494
Merck Index:14,08596