Potassium hydroxide 0.1 M in aqueous solution, Titrisol®, Supelco®
Catalog # CA1.09921.0001
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Cardboard box
- Size:1 pc.
- Environmentally Preferable:
- Cat. no.:CA1.09921.0001
- Supplier No.:1.09921.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.09 g/cm3 (20 °C)
- pH:14 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
Synonyms: Potash caustic, Caustic potash
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553