
Titriplex® solution B for the determination of alkaline earth in water (hardness) concentrate, Titrisol®, Supelco®
Catalog # CA1.09895.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:1 piece
- Cat. no.:CA1.09895.0001
- Supplier No.:1.09895.0001
Specifications
About this item
:
Density:1.10 g/cm3 (20 °C)
:GHS07
Storage Temperature:Ambient