Order Entry
Canada
ContactUsLinkComponent
Titrisol® Sulfate Standard, MilliporeSigma
Titrisol® Sulfate Standard, MilliporeSigma
Catalog # CA1.09872.0001
Supplier:  MilliporeSigma
CAS Number:  
Titrisol® Sulfate Standard, MilliporeSigma
Catalog # CA1.09872.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09872.0001
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Pack type:
    Cardboard box
  • Description:
    Sulfate
  • Size:
    1 pc
  • Cat. no.:
    CA1.09872.0001
  • Contents:
    H₂SO₄ in H₂O
  • Supplier No.:
    1.09872.0001

Specifications

About this item