Order Entry
Canada
ContactUsLinkComponent
Multi-Element Standard III Dissolved in Oil, Certipur®, MilliporeSigma
Multi-Element Standard III Dissolved in Oil, Certipur®, MilliporeSigma
Catalog # CA1.09479.0100
Supplier:  MilliporeSigma
CAS Number:  
Multi-Element Standard III Dissolved in Oil, Certipur®, MilliporeSigma
Catalog # CA1.09479.0100
Supplier:  MilliporeSigma
Supplier Number:  1.09479.0100
CAS Number:  

Specifications

  • Concentration:
    900 ppm
  • Pack type:
    Glass Bottle, Pre-Packed
  • Description:
    Multi-element standard III dissolved in oil
  • Size:
    100 ml
  • Cat. no.:
    CA1.09479.0100
  • Supplier No.:
    1.09479.0100

Specifications

About this item