To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Matrix:1% HNO₃
- Pack type:Plastic Bottle
- Description:ICP multi-element standard solution XXIV, for 700 ES (15 elements in dilute nitric acid) CertiPUR®
- Size:500 ml
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Elements and Concentration:50 mg/l: Al, As, Ba, Cd, Cr, Co, Cu, Pb, Mn, Mo, Ni, Se, Sr, Zn; 500 mg/l: K
- Cat. no.:CA1.09411.0500
- Supplier No.:1.09411.0500
Specifications
About this item
Element, Specification (mg/L), Uncertainty** (mg/L), NIST Standard Reference Material
Al, 45 - 55, ±5, SRM 3101a
As, 45 - 55, ±5, SRM 3103a
Ba, 45 - 55, ±5, SRM 3104a
Cd, 45 - 55, ±5, SRM 3108
Co, 45 - 55, ±5, SRM 3113
Cr, 45 - 55, ±5, SRM 3112a
Cu, 45 - 55, ±5, SRM 3114
K, 450 - 550, ±50, SRM 3141a
Mn, 45 - 55, ±5, SRM 3132
Mo, 45 - 55, ±5, SRM 3134
Ni, 45 - 55, ±5, SRM 3136
Pb, 45 - 55, ±5, SRM 3128
Se, 45 - 55, ±5, SRM 3149
Sr, 45 - 55, ±5, SRM 3153a
Zn, 45 - 55, ±5, SRM 3168a
**The measurement uncertainty can vary depending on the primary reference material.
Al, 45 - 55, ±5, SRM 3101a