Order Entry
Canada
ContactUsLinkComponent
Barium perchlorate 0.005 mol/l in 2-propanol/water (80/20), Titripur® Reag. USP volumetric solution, Supelco®
 
 
Barium perchlorate 0.005 mol/l in 2-propanol/water (80/20), Titripur® Reag. USP volumetric solution, Supelco®
  CA1.09086.1000
 :  
 
 
Barium perchlorate 0.005 mol/l in 2-propanol/water (80/20), Titripur® Reag. USP volumetric solution, Supelco®
  CA1.09086.1000
 :  1.09086.1000
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. no.:
    CA1.09086.1000
  • Supplier No.:
    1.09086.1000

 

 

  • Solubility:
    (20 °C) soluble
  • Flash Point:
    21 °C
  • Density:
    0.85 g/cm3 (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

 

 

 :
 :336.23 g/mol
 :3,II
 :M-Clarity™ quality level = MQ100
 :1993
 :GHS02
 :GHS07
 :13465-95-7
 :Ba(ClO₄)₂
 :MFCD00003440