Order Entry
Canada
ContactUsLinkComponent
 
 
 
Hydrochloric acid 3.571 mol/l (N/0.28), Titripur® volumetric solution, Supelco®
  CA1.13134.9010
 :  
 
 
Hydrochloric acid 3.571 mol/l (N/0.28), Titripur® volumetric solution, Supelco®
  CA1.13134.9010
 :  1.13134.9010
 :  

 

  • Size:
    10 L
  • Cat. no.:
    CA1.13134.9010
  • Supplier No.:
    1.13134.9010

 

 

  • Solubility:
    (20 °C) soluble
  • Density:
    1.02 g/cm3 (20 °C)
  • pH:
    \< - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

 

 :
 :1.06 g/cm³ (20 °C)
 :Ambient
 :231-595-7
 :1789
 :8,
 :M-Clarity™ quality level = MQ100
 :36.46
 :GHS05
 :GHS07
 :7647-01-0
 :HCl
 :MFCD00011324