Phenol red solution indicator, Supelco®
Supplier: MilliporeSigma
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Density:0.99 g/cm3 (20 °C)
- Storage Temperature:Ambient
1 Options Available Below
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Density:0.99 g/cm3 (20 °C)
- Storage Temperature:Ambient
Certificates
Search by Lot NumberSpecifications for Product Family
Specification Test Results
- Application test:conforms
- pH-value (20 °C):7.0 - 7.3
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Glass bottle100 mlDiscontinued-Supplier Number: 1072420100
- Glass bottle100 mlEach (100ml)$202.64Case of 15 (100ml)$3,039.60Supplier Number: 1.07242.0100
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



