Sodium carbonate, Certipur® volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
Catalog # CA1.02405.0080
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle, pre-packed
- Size:80 g
- Cat. no.:CA1.02405.0080
- Supplier No.:1.02405.0080
Specifications
Specification Test Results
- Solubility:220 g/l (20 °C)
- Melting Point:854 °C
- Molar Mass:105.99 g/mol
- Bulk Density:1100 kg/m3
- Boiling Point:1600 °C (decomposition)
- Density:2.53 g/cm3 (20 °C)
- pH:11.5 (50 g/l, H2O, 25 °C)
Specification Test Results
About this item
:
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494