Order Entry
Canada
ContactUsLinkComponent
Calcium chloride tetrahydrate, Suprapur® for trace metal analysis, Supelco®
Irritant
Calcium chloride tetrahydrate, Suprapur® for trace metal analysis, Supelco®
Catalog # CA1.02384.0100
Supplier:  MilliporeSigma
CAS Number:  
Irritant
Calcium chloride tetrahydrate, Suprapur® for trace metal analysis, Supelco®
Catalog # CA1.02384.0100
Supplier:  MilliporeSigma
Supplier Number:  1.02384.0100
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g
  • Cat. no.:
    CA1.02384.0100
  • Supplier No.:
    1.02384.0100

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    772 °C (anhydrous substance)
  • Molar Mass:
    183.05 g/mol
  • pH:
    5 - 7 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :183.04 g/mol
Boiling Pt:∼1600 °C (1013 hPa)
Melting Pt:772 °C
Density:1.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-140-8
 :GHS07
CAS Number:25094-02-4
Formula:CaCl₂·4H₂O