
Nitric acid 60%, Ultrapur for trace metal analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01
Boiling Pt:∼120 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-22 °C
- Boiling Point:120 °C (1013 hPa)
- Density:1.37 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Solutions for Sample Digestion Flyer MilliporeSigma High Purity Acids and Bases
Product Literature
ghs_1.01518
SDS
Ultrapur and Suprapur®High Purity Acid and Bases VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together