Order Entry
Canada
ContactUsLinkComponent
Water, Ultrapur for trace metal analysis, Supelco®
Water, Ultrapur for trace metal analysis, Supelco®
Catalog # CA1.01262.1000
Supplier:  MilliporeSigma
CAS Number:  
Water, Ultrapur for trace metal analysis, Supelco®
Catalog # CA1.01262.1000
Supplier:  MilliporeSigma
Supplier Number:  1.01262.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. no.:
    CA1.01262.1000
  • Supplier No.:
    1.01262.1000

Specifications

Specification Test Results

  • Ag (Silver):
    ≤ 50 ppt
  • Al (Aluminium):
    ≤ 200 ppt
  • As (Arsenic):
    ≤ 50 ppt
  • Au (Gold):
    ≤ 50 ppt
  • B (Boron):
    ≤ 500 ppt
  • Ba (Barium):
    ≤ 50 ppt
  • Be (Beryllium):
    ≤ 50 ppt
  • Bi (Bismuth):
    ≤ 50 ppt
  • Ca (Calcium):
    ≤ 500 ppt
  • Cd (Cadmium):
    ≤ 50 ppt
  • Ce (Cerium):
    ≤ 20.0 ppt
  • Co (Cobalt):
    ≤ 50 ppt
  • Cr (Chromium):
    ≤ 50 ppt
  • Cu (Copper):
    ≤ 100 ppt
  • Dy (Dysprosium):
    ≤ 20.0 ppt
  • Er (Erbium):
    ≤ 20.0 ppt
  • Eu (Europium):
    ≤ 20.0 ppt
  • Fe (Iron):
    ≤ 300 ppt
  • Ga (Gallium):
    ≤ 50.0 ppt
  • Gd (Gadolinium):
    ≤ 20.0 ppt
  • Ge (Germanium):
    ≤ 50 ppt
  • Hf(Hafnium):
    ≤ 10.0 ppt
  • Ho (Holmium):
    ≤ 20.0 ppt
  • In (Indium):
    ≤ 50.0 ppt
  • K (Potassium):
    ≤ 300 ppt
  • La (Lanthanum):
    ≤ 20.0 ppt
  • Li (Lithium):
    ≤ 50.0 ppt
  • Lu (Lutetium):
    ≤ 20.0 ppt
  • Mg (Magnesium):
    ≤ 100 ppt
  • Mn (Manganese):
    ≤ 50 ppt
  • Mo (Molybdenum):
    ≤ 50 ppt
  • Na (Sodium):
    ≤ 500 ppt
  • Nb (Niobium):
    ≤ 20.0 ppt
  • Nd (Neodymium):
    ≤ 20.0 ppt
  • Ni (Nickel):
    ≤ 50 ppt
  • Pb (Lead):
    ≤ 50 ppt
  • Pd (Palladium):
    ≤ 50 ppt
  • Pr (Praseodym):
    ≤ 20.0 ppt
  • Pt (Platinum):
    ≤ 50 ppt
  • Rb (Rubidium):
    ≤ 20.0 ppt
  • Sb (Antimony):
    ≤ 50 ppt
  • Sc (Scandium):
    ≤ 20 ppt
  • Si (Silicon):
    ≤ 1000 ppt
  • Sm (Samarium):
    ≤ 20.0 ppt
  • Sn (Tin):
    ≤ 50 ppt
  • Sr (Strontium):
    ≤ 50.0 ppt
  • Ta (Tantalum):
    ≤ 50.0 ppt
  • Tb (Terbium):
    ≤ 20.0 ppt
  • Te (Tellur):
    ≤ 50 ppt
  • Th (Thorium):
    ≤ 10.00 ppt
  • Ti (Titanium):
    ≤ 50 ppt
  • Tl (Thallium):
    ≤ 50.0 ppt
  • Tm (Thulium):
    ≤ 20.0 ppt
  • U (Uranium):
    ≤ 10.00 ppt
  • V (Vanadium):
    ≤ 50 ppt
  • W (Wolfram):
    ≤ 100 ppt
  • Y (Yttrium):
    ≤ 10.0 ppt
  • Yb (Ytterbium):
    ≤ 20.0 ppt
  • Zn (Zinc):
    ≤ 500 ppt
  • Zr (Zirconium):
    ≤ 50.0 ppt

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332