

Nitric acid 10 mol/l (10 N), Titripur® volumetric solution, Supelco®
Catalog # CA1.00630.1000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. no.:CA1.00630.1000
- Supplier No.:1.00630.1000
Specifications
Specification Test Results
- Assay (HNO₃):48.0 - 50.0 %
Specification Test Results
About this item
:
:63.01
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–28 °C
Density:1.38 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349