


Nitric acid 65%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01
Merck Index:13,06608
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.39 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8 (5.1),II
:M-Clarity™ quality level = MQ100
EINECS:231-714-2
:GHS03
:GHS06
:GHS05
Specification Test Results
- Assay (acidimetric):≥ 65.0 %
- Chloride (Cl):≤ 50 ppb
- Phosphate (PO₄):≤ 10 ppb
- Sulphate (SO₄):≤ 200 ppb
- Ag (Silver):≤ 0.5 ppb
- Al (Aluminium):≤ 10 ppb
- As (Arsenic):≤ 0.5 ppb
- Au (Gold):≤ 0.1 ppb
- Ba (Barium):≤ 0.5 ppb
- Be (Beryllium):≤ 0.5 ppb
- Bi (Bismuth):≤ 0.5 ppb
- Ca (Calcium):≤ 10 ppb
- Cd (Cadmium):≤ 0.5 ppb
- Co (Cobalt):≤ 0.5 ppb
- Cr (Chromium):≤ 1.0 ppb
- Cu (Copper):≤ 0.5 ppb
- Fe (Iron):≤ 2.0 ppb
- Ga (Gallium):≤ 0.1 ppb
- Ge (Germanium):≤ 0.1 ppb
- Hg (Mercury):≤ 1.0 ppb
- In (Indium):≤ 0.5 ppb
- K (Potassium):≤ 10 ppb
- Li (Lithium):≤ 0.5 ppb
- Mg (Magnesium):≤ 10 ppb
- Mn (Manganese):≤ 0.5 ppb
- Mo (Molybdenum):≤ 0.5 ppb
- Na (Sodium):≤ 10.0 ppb
- Ni (Nickel):≤ 5.0 ppb
- Pb (Lead):≤ 2.0 ppb
- Pt (Platinum):≤ 0.5 ppb
- Sb (Antimony):≤ 0.5 ppb
- Sn (Tin):≤ 0.5 ppb
- Sr (Strontium):≤ 0.5 ppb
- Ti (Titanium):≤ 1.0 ppb
- Tl (Thallium):≤ 0.5 ppb
- V (Vanadium):≤ 0.5 ppb
- Zn (Zinc):≤ 2.0 ppb
- Zr (Zirconium):≤ 1.0 ppb
- undefined:≤ 2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Solutions for Sample Digestion Flyer EMD Millipore Trace Elemental Analysis for USP 232-233 Brochure MilliporeSigma High Purity Acids and Bases
Product Literature
Ultrapur and Suprapur®High Purity Acid and Bases VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together